Iupac Name
5-fluoro-6-oxo-1,6-dihydro-3-pyridinecarboxylic acid
Inchi Code
1S/C6H4FNO3/c7-4-1-3(6(10)11)2-8-5(4)9/h1-2H,(H,8,9)(H,10,11)
InChI key
IEXXBKVOINSOFC-UHFFFAOYSA-N
storage temp.
Ambient Storage
Purity
95%
Country of Origin
GB
Product Link
https://fluorochem.co.uk/product/F550201
Signal Word
Warning
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/eng/sds_F550201_en.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service