Skip to Content
Merck

KEY465198677

Key Organics / BIONET

methyl 4-(4-chlorophenyl)-2-(2,4-dichlorophenyl)-4-oxobutanoate

Synonym(s):

methyl 4-(4-chlorophenyl)-2-(2,4-dichlorophenyl)-4-oxobutanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

500 MG

₹69,161.25

1 G

₹1,27,322.50

Vendor SKU: 7J-540S-500MG

₹69,161.25


Ships in 28 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
371.65
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C17H13Cl3O3/c1-23-17(22)14(13-7-6-12(19)8-15(13)20)9-16(21)10-2-4-11(18)5-3-10/h2-8,14H,9H2,1H3

Inchi Key

ZGKGKPUZGNXAHB-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Solid

MSDS

https://www.keyorganics.net/sds/7J-540S

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service