Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
41121811
Pricing and availability is not currently available.
manufacturer/tradename
BPS Bioscience
technique(s)
activity assay: suitable (Fluorescence Polarization)
UniProt accession no.
shipped in
dry ice
storage temp.
-65 to -96°C
General description
The PDE6C Assay Kit is a fluorescence polarization (FP), homogeneous, 96-well assay kit designed for the screening and profiling of PDE6C (Phosphodiesterase 6C) inhibitors. This assay takes advantage of a specific fluorescent phosphate-binding nanoparticle. The kit contains enough purified recombinant PDE6C, fluorescent probe, PDE assay buffer, Binding Agent, and diluent for 100 reactions. The assay uses a fluorescein-labeled cyclic guanosine monophosphate (cGMP-FAM for PDE6C), in which the phosphate group is engaged within the cyclic nucleotide. This is a very small molecule that rotates fast (low FP). PDE6C catalyzes the hydrolysis of the phosphodiester bond in the cyclic nucleotide and frees the phosphate group. In a second step the free phosphate group is recognized by a specific phosphate-binding nanobead (Binding Agent) leading to the formation of a large complex, with restricted movement (high FP). FP is proportional to PDE6C activity. This assay requires a fluorescent microplate reader capable of measuring fluorescence polarization (FP) and equipped with the required parts to read the FP signal.
Application
Study enzyme kinetics and screen small molecule inhibitors for drug discovery in high throughput screening (HTS) applications.
Phosphodiesterases (PDEs) play an important role in the dynamic regulation of the second messengers cAMP (cyclic adenosine monophosphate) and cGMP (cyclic guanosine monophosphate) signaling, by hydrolyzing them. The PDE superfamily is composed of 11 families, with PDE4, 7 and 8 being cAMP-specific hydrolases, and thus regulating positive and negative responses to it. PDE6C, also known as cGMP-inhibited phosphodiesterase, is involved in light detection and cone phototransduction. PDE6C encodes the alpha-prime subunit of cone-specific phosphodiesterase, an enzyme found exclusively in the light-detecting (photoreceptor) cells called cones. Cone cells are found in a specialized tissue at the back of the eye known as the retina, and mutations in PDE6c are associated with eye diseases including Cone Dystrophy 4 and Achromatopsia. Continuing to understand PDE6C roles and the development of newer specific therapeutic tools will open new treatment avenues for PDE-related disorders.
Datasheet and Protocols for the 96 reaction kit may be found here.
Datasheet and Protocols for the 384 reaction kit may be found here.
Phosphodiesterases (PDEs) play an important role in the dynamic regulation of the second messengers cAMP (cyclic adenosine monophosphate) and cGMP (cyclic guanosine monophosphate) signaling, by hydrolyzing them. The PDE superfamily is composed of 11 families, with PDE4, 7 and 8 being cAMP-specific hydrolases, and thus regulating positive and negative responses to it. PDE6C, also known as cGMP-inhibited phosphodiesterase, is involved in light detection and cone phototransduction. PDE6C encodes the alpha-prime subunit of cone-specific phosphodiesterase, an enzyme found exclusively in the light-detecting (photoreceptor) cells called cones. Cone cells are found in a specialized tissue at the back of the eye known as the retina, and mutations in PDE6c are associated with eye diseases including Cone Dystrophy 4 and Achromatopsia. Continuing to understand PDE6C roles and the development of newer specific therapeutic tools will open new treatment avenues for PDE-related disorders.
Datasheet and Protocols for the 96 reaction kit may be found here.
Datasheet and Protocols for the 384 reaction kit may be found here.
Features and Benefits
The PDE6C inhibitor screening assay kit comes in a convenient 96-well format, with purified PDE6C enzyme, fluorescently labeled PDE6 substrate (cGMP), binding agent, and PDE assay buffer for 100 enzyme reactions
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
