Merck
  • GTS-21 Promotes α7 nAChR to Alleviate Intestinal Ischemia-Reperfusion-Induced Apoptosis and Inflammation of Enterocytes.
GTS-21 Promotes α7 nAChR to Alleviate Intestinal Ischemia-Reperfusion-Induced Apoptosis and Inflammation of Enterocytes.
Medical science monitor : international medical journal of experimental and clinical research (2020-05-18)
Haisong Wang, Dongmiao Cai, Zhenyi Chen, Yanlin Wang
ABSTRACT
BACKGROUND Intestinal ischemia-reperfusion injury is a serious intestinal disease, with main symptoms of inflammatory reaction and severe oxidative damage. In addition, GTS-21-induced alpha7 nAChR has been shown to exert anti-inflammatory effects and anti-oxidation effects in various organs. However, whether alpha7 nAChR can alleviate ischemia-reperfusion-induced intestinal injury is unclear. MATERIAL AND METHODS We used intestinal epithelial cells (IEC-6) to perform the experiments. Oxygen glucose deprivation/reoxygenation (OGD/R) was used to simulate the physiological environment of ischemia-reperfusion. First, the expression of alpha7 nAChR was determined in these cells which was cultured under OGD/R conditions. After that, the GTS-21 was used to treat these cells and the levels of inflammatory factors (TNF-alpha, IL-1ß, IL-6, and IL-10) were assessed by ELISA. Next, the levels of ROS, SOD, and MDA were determined in IEC-6 cells. Finally, the apoptosis rates of IEC-6 cells were measured by flow cytometry. RESULTS Results showed that the expression of TNF-alpha, IL-1ß, and IL-6 was enhanced when the IEC-6 cells were cultured under OGD/R conditions. However, after treatment with GTS-21, the levels of these proinflammatory factors were suppressed. In addition, the levels of ROS and MDA were also inhibited and the expression of SOD was promoted after GTS-21 treatment. We also found that the ratios of apoptotic cells declined after GTS-21 treatment. CONCLUSIONS GTS-21-induced alpha7 nAChR decreased the OGD/R-induced inflammatory response, oxidative damage, and apoptosis of intestinal epithelial cells.
Sigma-Aldrich
Human IL-1 β ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
IL-10 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)