Merck
  • Late onset of treatment with a chemokine receptor CCR1 antagonist prevents progression of lupus nephritis in MRL-Fas(lpr) mice.
Late onset of treatment with a chemokine receptor CCR1 antagonist prevents progression of lupus nephritis in MRL-Fas(lpr) mice.
Journal of the American Society of Nephrology : JASN (2004-05-22)
Hans-Joachim Anders, Emilia Belemezova, Vaclav Eis, Stephan Segerer, Volker Vielhauer, Guillermo Perez de Lema, Matthias Kretzler, Clemens D Cohen, Michael Frink, Richard Horuk, Kelly L Hudkins, Charles E Alpers, Francisco Mampaso, Detlef Schlöndorff
ABSTRACT
Slowly progressive renal injury is the major cause for ESRD. The model of progressive immune complex glomerulonephritis in autoimmune MRL(lpr/lpr) mice was used to evaluate whether chemokine receptor CCR1 blockade late in the disease course can affect progression to renal failure. Mice were treated with subcutaneous injections of either vehicle or BX471, a nonpeptide CCR1 antagonist, three times a day from week 20 to 24 of age [corrected]. BX471 improved blood urea nitrogen levels (BX471, 35.1 +/- 5.3; vehicle, 73.1 +/- 39.6 mg/dl; P < 0.05) and reduced the amount of ERHR-3 macrophages, CD3 lymphocytes, Ki-67 positive proliferating cells, and ssDNA positive apoptotic cells in the interstitium but not in glomeruli. Cell transfer studies with fluorescence-labeled T cells that were pretreated with either vehicle or BX471 showed that BX471 blocks macrophage and T cell recruitment to the renal interstitium of MRL(lpr/lpr) mice. This was associated with reduced renal expression of CC chemokines CCL2, CCL3, CCL4, and CCL5 and the chemokine receptors CCR1, CCR2, and CCR5. Furthermore, BX471 reduced the extent of interstitial fibrosis as evaluated by interstitial smooth muscle actin expression and collagen I deposits, as well as mRNA expression for collagen I and TGF-beta. BX471 did not affect serum DNA autoantibodies, proteinuria, or markers of glomerular injury in MRL(lpr/lpr) mice. This is the first evidence that, in advanced chronic renal injury, blockade of CCR1 can halt disease progression and improve renal function by selective inhibition of interstitial leukocyte recruitment and fibrosis.
Sigma-Aldrich
BX471, ≥98% (HPLC)
Pricing and availability is not currently available.