Merck
  • Fluazinam targets mitochondrial complex I to induce reactive oxygen species-dependent cytotoxicity in SH-SY5Y cells.
Fluazinam targets mitochondrial complex I to induce reactive oxygen species-dependent cytotoxicity in SH-SY5Y cells.
Neurochemistry international (2012-04-03)
Jeong Eun Lee, Jin Sun Kang, Yeo-Woon Ki, Jae Hyeon Park, In Chul Shin, Hyun Chul Koh
ABSTRACT
Although the underlying cause of Parkinson's disease (PD) is not well characterized, epidemiological studies suggest that exposure to agricultural chemicals is a risk factor for PD. Fluazinam (FZN) is a new active ingredient for the control of grey mould, belonging to the novel broad spectrum phenylpyridinamine fungicides. We used human neuroblastoma SH-SY5Y cells to investigate mechanisms of dopaminergic cell death in response to FZN. FZN treatment produced dose-dependent cytotoxicity, and decreased the tyrosine hydroxylase (TH) expression in SH-SY5Y cells. We provided evidence for the occurrence of oxidative stress and oxidative damage during FZN exposure on dopaminergic cells through the measurement of reactive oxygen species (ROS) in cells with DCFH-DA. The cytotoxic effects of FZN appear to involve an increase in ROS generation since pretreatment with N-acetyl cysteine (NAC), an anti-oxidant, reduced cell death. After FZN treatment, dopamine (DA) levels decreased in both cell and culture media, and oxidative effects of FZN were blocked by NAC pretreatment. We show that cell death in response to FZN was due to apoptosis since FZN exposure results in an increased in cytochrome c release into the cytosol and activated caspase-3 through p38 and JNK signaling. Furthermore, the blocking of p38 or JNK signaling inhibits FZN-induced cell death. Phosphorylation of mitogen-activated protein kinases precedes cytochrome c release and caspase-3 activation. This cellular response is characteristic of mitochondrial dysfunction. Therefore, we also investigated the effect of FZN on mitochondrial complex I activity in FZN-treated cell. Interestingly, we show that FZN inhibited the complex I activity. Thus in this study, we report a new mode of action by which the fungicide FZN could triggers apoptosis.
Product Description
Supelco
Fluazinam, PESTANAL®, analytical standard
Pricing and availability is not currently available.