Merck
  • Matrix metalloproteinase activity correlates with blastema formation in the regenerating MRL mouse ear hole model.
Matrix metalloproteinase activity correlates with blastema formation in the regenerating MRL mouse ear hole model.
Developmental dynamics : an official publication of the American Association of Anatomists (2003-01-31)
Dmitri Gourevitch, Lise Clark, Pan Chen, Alexander Seitz, Stefan J Samulewicz, Ellen Heber-Katz
ABSTRACT
The MRL mouse was proposed as a model of mammalian regeneration because it can close ear holes completely with the restoration of normal tissue. This regeneration process involves the formation of a blastema during healing, the re-appearance of cartilage and hair follicles, and healing without scarring. Such a process requires extensive tissue remodeling. To characterize differences in ear wounding responses between regenerating and nonregenerating mice, we examined and compared the extracellular matrix remodeling and the matrix metalloproteinase (MMP) and tissue inhibitor of metalloproteinase (TIMP) response in the MRL and C57BL/6 mouse strains after injury. We found a correlation between the MRL's ability to break down the basement membrane, form a blastema, and close ear hole wounds and an inflammatory response with neutrophils and macrophages seen in the ear after injury. These cells were positive for MMP-2 and MMP-9 as well as TIMP-2 and TIMP-3. Clear differences between the MRL and B6 response to injury were seen that could explain the differences in healing and blastema formation in the MRL and lack of it in the B6 mice. This finding was further supported by enzyme activity as determined by gelatin zymography.