Merck
  • Specific Ser-Pro phosphorylation by the RNA-recognition motif containing kinase KIS.
Specific Ser-Pro phosphorylation by the RNA-recognition motif containing kinase KIS.
European journal of biochemistry (2000-07-06)
A Maucuer, J P Le Caer, V Manceau, A Sobel
ABSTRACT
We present here a first appraisal of the phosphorylation site specificity of KIS (for 'kinase interacting with stathmin'), a novel mammalian kinase that has the unique feature among kinases to possess an RNP type RNA-recognition motif (RRM). In vitro kinase assays using various standard substrates revealed that KIS has a narrow specificity, with myelin basic protein (MBP) and synapsin I being the best in vitro substrates among those tested. Mass spectrometry and peptide sequencing allowed us to identify serine 164 of MBP as the unique site phosphorylated by KIS. Phosphorylation of synthetic peptides indicated the importance of the proline residue at position +1. We also identified a tryptic peptide of synapsin I phosphorylated by KIS and containing a phosphorylatable Ser-Pro motif. Altogether, our results suggest that KIS preferentially phosphorylates proline directed residues but has a specificity different from that of MAP kinases and cdks.
Sigma-Aldrich
Myelin Basic Protein bovine, ≥90% (SDS-PAGE), lyophilized powder, suitable for activity assay
Sigma-Aldrich
Histone from calf thymus, Type III-S