Merck
  • Adenosine A2A receptor signaling affects IL-21/IL-22 cytokines and GATA3/T-bet transcription factor expression in CD4
Adenosine A2A receptor signaling affects IL-21/IL-22 cytokines and GATA3/T-bet transcription factor expression in CD4
Journal of neuroimmunology (2017-08-16)
Sheikh F Ahmad, Mushtaq A Ansari, Ahmed Nadeem, Saleh A Bakheet, Mashal M Almutairi, Sabry M Attia
ABSTRACT
Autism is a complex heterogeneous neurodevelopmental disorder; previous studies have identified altered immune responses among individuals diagnosed with autism. An imbalance in the production of pro- and anti-inflammatory cytokines and transcription factors plays a role in neurodevelopmental behavioral and autism disorders. BTBR T
Sigma-Aldrich
SCH 58261, ≥98% (HPLC), solid
Pricing and availability is not currently available.