Merck
Consolidation of the optoelectronic properties of CH
Nature communications (2017-09-21)
Bernard Wenger, Pabitra K Nayak, Xiaoming Wen, Sameer V Kesava, Nakita K Noel, Henry J Snaith
ABSTRACT
Ultralow trap densities, exceptional optical and electronic properties have been reported for lead halide perovskites single crystals; however, ambiguities in basic properties, such as the band gap, and the electronic defect densities in the bulk and at the surface prevail. Here, we synthesize single crystals of methylammonium lead bromide (CH
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Pricing and availability is not currently available.