- GAK: a cyclin G associated kinase contains a tensin/auxilin-like domain.
GAK: a cyclin G associated kinase contains a tensin/auxilin-like domain.
FEBS letters (1997-01-27)
Y Kanaoka, S H Kimura, I Okazaki, M Ikeda, H Nojima
PMID9013862
ABSTRACT
We have cloned a cDNA encoding a novel association partner of cyclin G by West-Western blotting. The cDNA encodes a protein that harbors a Ser/Thr protein kinase-like catalytic domain at the N-terminal. Hence, we named it GAK (cyclin G-associated kinase). The long C-terminal extension shares homology with tensin and auxilin, and contains a leucine zipper region. Co-immunoprecipitation and Western blotting showed that GAK and cyclin G associate together in vivo. GAK also co-precipitated with CDK5, and CDK5 was found to be associated with cyclin G. We also showed by BIAcore analysis that the GAK-cyclin G interaction was direct.