Merck
LC/MS Analysis of Sudan Red Dyes on Ascentis® RP-Amide
LC/MS Analysis of Sudan Red Dyes on Ascentis® RP-Amide application for HPLC
Materials
related product
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis RP-Amide, 15 cm × 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
[A] 0.1% formic acid in water (pH unadjusted): [B] acetonitrile; (15:85, A:B)
flow rate
1 mL/min, split to the MS
column temp.
35 °C
detector
MS, APCI (+) in selected ion recording (SIR) mode
injection
15 μL
sample
1 μg/mL in 30:70, 0.1% formic acid in water (pH unadjusted):acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany