Merck
LC/MS/MS Analysis of Multiple Mycotoxins on Ascentis® Express Phenyl-Hexyl
LC/MS/MS Analysis of Multiple Mycotoxins on Ascentis® Express Phenyl-Hexyl application for LC-MS
Materials
related product
Product No.
Description
Pricing
Deoxynivalenol solution
100 μg/mL in acetonitrile, analytical standard
Fumonisin Mixture
~50 μg/mL in acetonitrile: water (each of FB1 and FB2), analytical standard
Fumonisin B1
reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
15-Acetyldeoxynivalenol
reference material
3-Acetyldeoxynivalenol
Sterigmatocystin solution
~50 μg/mL in acetonitrile, analytical standard
Ascentis® Express Phenyl-Hexyl (2.7 μm) HPLC Column
L × I.D. 10 cm × 2.1 mm, pk of 1 ea
Citrinin solution
~100 μg/mL in acetonitrile, analytical standard
Aflatoxin Mix
certified reference material, in methanol (varied conc.), ampule of 5 × 1 mL
Ochratoxin A solution
certified reference material, 50 μg/mL in benzene: acetic acid (99:1), ampule of 1 mL
Zearalenone solution
certified reference material, 50 μg/mL in acetonitrile, ampule of 1 mL
CONDITIONS
column
Ascentis Express Phenyl-Hexyl, 10 cm x 2.1 mm I.D., 2.7 μm particles (53336-U)
mobile phase
[A] 5mM ammonium formate with 0.1% formic acid in water ; [B] 5mM ammonium formate with 0.1% formic acid in acetonitrile
gradient
30 to 60% B in 3 min; to 100% B in 2 min; hold at 100% B for 2 min; 100 to 30% B in 0.5 min; hold at 30% for 2 min
flow rate
0.4 mL/min
pressure
5511 psi (380 bar)
column temp.
40 °C
detector
MS/MS, ESI(+), MRM (See Figure for transitions)
injection
2 μL
Description
Analysis Note
Fast and selective method for 16 mycotoxins.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany