Skip to Content
Merck

560464

3-(4-Morpholino)propyl isothiocyanate

95%

Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C8H14N2OS
CAS Number:
Molecular Weight:
186.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

95%

refractive index

n20/D 1.5303 (lit.)

density

1.094 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C=NCCCN1CCOCC1

InChI

1S/C8H14N2OS/c12-8-9-2-1-3-10-4-6-11-7-5-10/h1-7H2

InChI key

BCEFDMYFAAAFPE-UHFFFAOYSA-N

Application

3-(4-Morpholino)propyl isothiocyanate may be used to synthesize (R,S)-(3-morpholin-4-yl-propyl)-thiocarbamic acid O-(5-[1,2] dithiolan-3-yl-pentyl) ester hydrochloride[1] and N-(3-N,N-dimethylaminopropyl-N′-3-(4-morpholino)propylthiourea.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claude Guillonneau et al.
European journal of medicinal chemistry, 38(1), 1-11 (2003-02-21)
Molecules containing a dithiolane moiety are widely investigated due to their antioxidant properties. The archetypal representative of this class of compounds is lipoic acid and indeed the lipoic acid-dihydrolipoic acid couple is part of the antioxidant defence system of the
Synthesis of Novel Fluorescent Reagents with a Carbodiimide Moiety and Their Applications for DNA Probe.
Tachikawa T, et al.
J. Photopolym. Sci. Technol., 11(1), 19-22 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service