Skip to Content
Merck

BDPH9BCC89C8

Bide Pharmatech

5-Chloronicotinic acid

Synonym(s):

5-chloronicotinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4O2N1Cl1
CAS Number:
Molecular Weight:
157.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H4ClNO2/c7-5-1-4(6(9)10)2-8-3-5/h1-3H,(H,9,10)

Inchi Key

XYLPLVUYPAPCML-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Very pale yellow



Still not finding the right product?

Explore all of our products under 5-Chloronicotinic acid


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library