Skip to Content
Merck

BDPH9BCCAB2F

Bide Pharmatech

2-Fluoro-5-methoxyphenol

Synonym(s):

2-fluoro-5-methoxyphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O2F1
CAS Number:
Molecular Weight:
142.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7FO2/c1-10-5-2-3-6(8)7(9)4-5/h2-4,9H,1H3

Inchi Key

KGCLSTPUECTXNO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid



Still not finding the right product?

Explore all of our products under 2-Fluoro-5-methoxyphenol


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library