Skip to Content
Merck

BDPH9BCD45FB

Bide Pharmatech

1-Boc-3-(2-aminoethyl)piperidine

Synonym(s):

tert-butyl 3-(2-aminoethyl)-1-piperidinecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H24O2N2
CAS Number:
Molecular Weight:
228.33
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H24N2O2/c1-12(2,3)16-11(15)14-8-4-5-10(9-14)6-7-13/h10H,4-9,13H2,1-3H3

Inchi Key

GYLAYRXNMUUXJS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to pale-yellow liquid



Still not finding the right product?

Explore all of our products under 1-Boc-3-(2-aminoethyl)piperidine


Precautionary Statements

Signal Word

Danger

Pictograms

CorrosionExclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library