Skip to Content
Merck

BDPH9BCDFAA0

Bide Pharmatech

2-Bromo-3-(bromomethyl)pyridine

Synonym(s):

2-bromo-3-(bromomethyl)pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5N1Br2
CAS Number:
Molecular Weight:
250.92
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H5Br2N/c7-4-5-2-1-3-9-6(5)8/h1-3H,4H2

Inchi Key

NEWLOSCWMVKPGI-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid or Semi-Solid or Liquid or Lump



Still not finding the right product?

Explore all of our products under 2-Bromo-3-(bromomethyl)pyridine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library