Skip to Content
Merck

BDPH9C021A07

Bide Pharmatech

2-Chloro-N-(cyanomethyl)acetamide

Synonym(s):

2-chloro-N-(cyanomethyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H5O1N2Cl1
CAS Number:
Molecular Weight:
132.55
MDL number:
Product BDPH9C021A07 is not currently sold in your country.


Inchi Code

1S/C4H5ClN2O/c5-3-4(8)7-2-1-6/h2-3H2,(H,7,8)

Inchi Key

BLGRCGIMBCJLMZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 2-Chloro-N-(cyanomethyl)acetamide



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library