Skip to Content
Merck

14-522-M

ABL (T315I) Protein, active

recombinant, expressed in baculovirus infected Sf21 cells

Sign In to View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Abl (T315I) Protein, active, 10 µg, Active, N-terminal His6-tagged recombinant human Abl, residues 27-end containing the T315I mutation. For use in Kinase Assays.

biological source

human

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 121 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal His6-tagged recombinant human Abl, residues 27-end containing the T315I mutation

Contains the gatekeeper mutation T315I in the ATP-binding domain. This mutation decreases the sensitivity to Gleevec by more than 200-fold as compared to the wild type sequence.
Product Source: Expressed by baculovirus in Sf21 insect cells

Application

Research Category
Metabolism

Inflammation & Immunology
Research Sub Category
Obesity

Metabolic Disorders

Osteoporosis

Arthritis

Biochem/physiol Actions

Protein Target: Abl
Target Sub-Family: TK

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-522M when ordering the 250 µg size.

Physical form

Ni2+/NTA-agarose

Preparation Note

6 months at -70°C

Analysis Note

routinely evaluated by phosphorylation of Abltide

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A non-ATP-competitive inhibitor of BCR-ABL overrides imatinib resistance
Gumireddy, K., et al
Proceedings of the National Academy of Sciences of the USA, 102, 1992-1997 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service