Skip to Content
Merck

5.30153

JAK2 Inhibitor X, FLLL32

Synonym(s):

JAK2 Inhibitor X, FLLL32, STAT3 Signaling Inhibitor, FLLL32

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C28H32O6
CAS Number:
Molecular Weight:
464.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

5 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell permeable diketone analog of curcumin that acts as a specific inhibitor of JAK2 (~ 5 µM) and blocks STAT3 phosphorylation in MDA-MB-231 breast and PANC-1 pancreatic cancer cells. However, it does not affect STAT3 phosphorylation in normal human mammary epithelial cells or human pancreatic duct epithelial cells. Does not block the activity of tyrosine kinases containing SH2 and SH2-SH3 domains and several other kinases, such as Akt2, Cdk2-cyclin D1, EGFR kinase, Met, and EbrB2/Her2 kinase (IC50 >100 µM). Also shown to block DNA binding activity of STAT3. Significantly reduces the viability and invasion capacity of various cancer cells (IC50 = 640 nM; 110 nM, 140 nM, 600 nM, 400 nM in PANC-1, HPAC, MDA-MB-231, SK-BR3, and SUM-159 cells, respectively) and induces apoptosis. Shown to significantly reduce tumor burdens in the MDA-MB-231 xenografted mice.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
JAK2
Reversible: yes

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wu, J., et al. 2014. Oncogene33,173.
Onimoe, G., et al. 2012. Invest. New Drugs30, 916.
Li, L., et al. 2010. Cancer Res.70, 2445.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service