Skip to Content
Merck

ABT108

Anti-Lnk/SH2B adapter protein 3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SH2B adapter protein 3, Lymphocyte adapter protein, Lymphocyte-specific adapter protein Lnk, Signal transduction protein Lnk

Sign In to View Organizational & Contract Pricing

Select a Size

100 μG

€348.00

€348.00


Estimated to ship onDecember 02, 2025Details


Request a Bulk Order

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SH2B3(10019)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA005483SAB1300534MABS1178
Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

technique(s)

western blot: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL

technique(s)

immunohistochemistry: 1:50-1:100, indirect ELISA: 1:1000

technique(s)

western blot: suitable

Gene Information

human ... SH2B3(10019)

Gene Information

human ... SH2B3(10019)

Gene Information

human ... SHB(6461)

Gene Information

human ... SH2B3(10019)

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

shipped in

ambient

General description

Lnk is an SH2B3 multidomain adaptor protein that is primarily expressed in lymphocytes. It contains the characteristic pleckstrin homology (PH); src homology 2 (SH2); dimerization; and proline-rich domains. Lnk functions as a negative regulator of B cell proliferative processes via interaction with Jak2 and Mpl proteins, and inhibition of the PI3/Akt/STAT5 signaling pathway. Previous studies have implicated Lnk in celiac disease type 13.
~80 kDa observed. The calculated molecular weight is 63 kDa, however Lnk/SH2B adapter protein 3 has been shown as a ~80 kDa band in western blots (Li, Y., et al. (2000). J Immunol. 164:5199-5206.)

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human Lnk/SH2B adapter protein 3.

Application

Detect the Lnk/SH2B adapter protein 3 protein using this Anti-Lnk/SH2B adapter protein 3 validated for use in WB.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Biochem/physiol Actions

Other homologies: Rat (78% sequence homology) and Mouse (72% sequence homology).
This antibody recognizes Lnk/SH2B adapter protein 3 at the C-terminus.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate
Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Lnk/SH2B adapter protein 3 on 10 µg of HepG2 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

New Products: Antibodies, Assays, Small Molecules, Inhibitors, and Proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service