Skip to Content
Merck

51530

Heneicosanoic acid

≥98.0% (GC)

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Linear Formula:
CH3(CH2)19COOH
CAS Number:
Molecular Weight:
326.56
Beilstein:
1711889
EC Number:
MDL number:
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

≥98.0% (GC)

mp

74-75 °C (lit.)
74-76 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C21H42O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23/h2-20H2,1H3,(H,22,23)

InChI key

CKDDRHZIAZRDBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A V Sarma et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 19(4), 671-676 (2001-08-24)
The mechanism of lubrication in normal human joints depends on loading and velocity conditions. Boundary lubrication, a mechanism in which layers of molecules separate opposing surfaces, occurs under severe loading. This study was aimed at characterizing the phospholipid composition of
Alexandre G Torres et al.
The British journal of nutrition, 95(3), 517-524 (2006-03-04)
Maternal adipose tissue is a major contributor to breast milk long-chain fatty acids, probably through the pool of plasma NEFA. The fatty acid composition of the erythrocyte membrane (EM) is a biochemical index of the intake of fatty acids not
Erin E Mosley et al.
The American journal of clinical nutrition, 82(6), 1292-1297 (2005-12-08)
trans Fatty acids (FAs) have been identified as negatively affecting human health. The trans FA composition of human milk fat must be examined to establish its influence on the nutritional quality of milk consumed by infants. We sought to ascertain
Olga Aksanov et al.
Cellular and molecular life sciences : CMLS, 71(17), 3381-3392 (2014-02-07)
BBS4 is one of several proteins whose defects cause Bardet-Biedl syndrome (BBS), a multi-systemic disorder, manifesting with marked obesity. BBS4 polymorphisms have been associated with common non-syndromic morbid obesity. BBS4 obesity molecular mechanisms, and the role of the BBS4 gene
Yunbao Liu et al.
Journal of natural products, 73(7), 1193-1195 (2010-06-23)
Antioxidants scavenge free radicals, singlet oxygen, and electrons in cellular redox reactions. The yellow MTT [3-(4,5-dimethylthiazole-2-yl)-2,5-diphenyltetrazolium bromide] is reduced to a purple formazan by mitochondrial enzymes. NADPH is the basis of established in vitro cell viability assays. An antioxidant assay

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service