Select a Size
€726.00
€726.00
About This Item
Skip To
biological source
human
recombinant
expressed in baculovirus infected insect cells
Assay
≥70% (SDS-PAGE)
form
aqueous solution
mol wt
51 kDa (EED)
86 kDa (EZH2)
packaging
pkg of 20 μg
concentration
>0.02 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... EZH2(2146)
1 of 4
This Item | SRP0132 | SRP0130 | SRP0131 |
|---|---|---|---|
| assay ≥70% (SDS-PAGE) | assay ≥90% (SDS-PAGE) | assay ≥80% (SDS-PAGE) | assay ≥90% (SDS-PAGE) |
| biological source human | biological source human | biological source human | biological source human |
| recombinant expressed in baculovirus infected insect cells | recombinant expressed in baculovirus infected insect cells | recombinant expressed in baculovirus infected insect cells | recombinant expressed in baculovirus infected insect cells |
| concentration >0.02 mg/mL | concentration >0.02 mg/mL | concentration >0.02 mg/mL | concentration >0.02 mg/mL |
| mol wt 51 kDa (EED), 86 kDa (EZH2) | mol wt 51 kDa (EED), 86 kDa (EZH2), 87 kDa (SUZ12) | mol wt 51 kDa (EED), 86 kDa (EZH1), 87 kDa (SUZ12) | mol wt 48 kDa (RbAp48), 51 kDa (EED), 53 kDa (AEBP2), 86 kDa (EZH1), 87 kDa (SUZ12) |
| form aqueous solution | form aqueous solution | form aqueous solution | form aqueous solution |
General description
Application
Physical form
Preparation Note
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


