Merck
  • Suppression of puerarin on polymethylmethacrylate-induced lesion of peri-implant by inhibiting NF-κB activation in vitro and in vivo.
Suppression of puerarin on polymethylmethacrylate-induced lesion of peri-implant by inhibiting NF-κB activation in vitro and in vivo.
Pathology, research and practice (2019-03-12)
Bo Yan, Hong Zhou, Jinpu Chu, Xuanping Cao
ABSTRACT
Puerarin (PR), a natural isoflavone isolated from Chinese traditional plant pueraria lobata, has attracted considerable attention due to its important biological and pharmacological activities. However, its effects on lesion of peri-implant and related mechanism of action are still not clear, which require further investigation. In this study, we evaluated the effects of PR on polymethylmethacrylate (PMMA)-induced lesion of peri-implant in vitro and in vivo, and explored its possible mechanism of action. Our results indicated that PR could inhibit PMMA-induced osteoclastogenesis in RAW264.7 cells with a dose-dependent manner in vitro and effectively down-regulate mRNA and protein expressions of matrix metalloprotein 9 (MMP-9), tumor necrosis factor (TNF)-α, interleukin (IL)-6, and receptor activator of nuclear factor (NF)-κB (RANK), primarily via the suppression of NF-κB signaling. Furthermore, we found that PMMA induction could directly cause the phosphorylation of IκB and significantly promote the nuclear translocation of p65 in RAW264.7 cells. In other words, PR was able to dose-dependently attenuate the PMMA-induced nuclear translocation of p65 in RAW264.7 cells. In vivo, PR was observed to attenuate PMMA-induced osteoclastogenesis, osteolysis, mRNA expressions of receptor activator of nuclear factor (NF)-κB ligand (RANKL) and RANK, as well as protein levels of MMP-9, TNF-α, IL-6, and p65 in a murine calvarial osteolysis model. These findings suggested that PR might be a potential therapeutic drug to lesion of peri-implant, and provided new insights for understanding its possible mechanism.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)