Merck
  • Does the Addition of Abiraterone to Castration Affect the Reduction in Bone Mineral Density?
Does the Addition of Abiraterone to Castration Affect the Reduction in Bone Mineral Density?
In vivo (Athens, Greece) (2020-11-05)
Shiori Nakajima, Takamitsu Inoue, Mingguo Huang, Koichiro Takayama, Soki Kashima, Ryohei Yamamoto, Atsushi Koizumi, Taketoshi Nara, Kazuyuki Numakura, Mitsuru Saito, Shintaro Narita, Masatomo Miura, Shigeru Satoh, Tomonori Habuchi
ABSTRACT
The in vivo effect of abiraterone on bone mineral density (BMD) in addition to androgen deprivation therapy was examined using a murine model. The mice were separated into the following groups: control, abiraterone, castration, and castration+abiraterone. The percentage change in the ratio of bone to tissue volume (BV/TV), number of osteoblasts and osteoclasts, and the serum level of bone markers were compared on day 21. The BV/TV ratio of the abiraterone, castration, and castration+abiraterone groups was lower than that of the control group. However, the change in the BV/TV ratio in the castration+abiraterone group was not significantly different from that in the castration group. There was no significant difference in the serum TRAP5b level and the number of osteoclasts and osteoblasts between the castration+abiraterone and the castration groups. The addition of abiraterone to castration did not affect BMD in the murine model.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Human TNFRSF11A ELISA Kit, for serum, plasma, cell culture supernatants and urine