Merck
  • Differential activation and redistribution of c-Src and Fyn in platelets, assessed by MoAb specific for C-terminal tyrosine-dephosphorylated c-Src and Fyn.
Differential activation and redistribution of c-Src and Fyn in platelets, assessed by MoAb specific for C-terminal tyrosine-dephosphorylated c-Src and Fyn.
Biochimica et biophysica acta (2000-06-06)
Y Wu, Y Ozaki, K Inoue, K Satoh, T Ohmori, Y Yatomi, K Owadab
ABSTRACT
Tyrosine kinases, c-Src and Fyn, in their active form, have their C-terminal tyrosine residue dephosphorylated. In this study, we used clone 28, a monoclonal antibody (MoAb) that recognizes dephosphorylated C-terminal tyrosine of c-Src and Fyn, to investigate the mode of activation and mobilization of these kinases. Independently of integrin alphaIIbbeta3 signaling, the Fyn activity increased by 8.3-fold 5 s after stimulation with 20 microM TRAP (thrombin receptor agonist peptide), while that of c-Src increased only by 2.9-fold 15 s after stimulation. Both c-Src and Fyn translocated to the Triton-insoluble cytoskeletal fraction in an aggregation-dependent manner. Five minutes after TRAP-stimulation, 85% of Fyn translocated to the cytoskeleton, while only about 20% of c-Src was recovered in this fraction. The Triton-insoluble fraction was further fractionated by RIPA (radioimmunoprecipitation assay) buffer containing 0.1% SDS. While active c-Src was predominantly present in the Triton-insoluble/RIPA-insoluble fraction, clone 28-negative c-Src was present in the Triton-insoluble/RIPA-soluble fraction. On the other hand, Fyn was present only in the Triton-insoluble/RIPA-insoluble fraction. These findings suggest that the mode of activation and redistribution into the cytoskeleton differs between c-Src and Fyn, and that clone 28 provides a useful tool for investigating the activation and mobilization of Src family tyrosine kinases.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)