Merck
  • Development of novel sol-gel coatings by chemically bonded ionic liquids for stir bar sorptive extraction--application for the determination of NSAIDS in real samples.
Development of novel sol-gel coatings by chemically bonded ionic liquids for stir bar sorptive extraction--application for the determination of NSAIDS in real samples.
Analytical and bioanalytical chemistry (2014-09-23)
Wenying Fan, Xiangju Mao, Man He, Beibei Chen, Bin Hu
ABSTRACT
In this work, a novel ionic liquid (IL) chemically bonded sol-gel coating was prepared for stir bar sorptive extraction (SBSE) of nonsteroidal anti-inflammatory drugs (NSAIDs) followed by high-performance liquid chromatography-ultraviolet detection (HPLC-UV). By using γ-(methacryloxypropyl)trimethoxysilane (KH-570) as a bridging agent, 1-allylimidazolium tetrafluoroborate ([AIM][BF4]) was chemically bonded onto the bare stir bar, and the prepared IL-bonded sol-gel stir bar coating showed higher extraction efficiency and better adsorption/desorption kinetics for target NSAIDs over other polydimethylsiloxane (PDMS)-based or monolithic stir bar coatings. The mechanical strength and durability (chemical/thermal stability) of the prepared IL-bonded sol-gel coating were excellent. The influencing factors of SBSE, such as sample pH, salt effect, stirring rate, extraction time, desorption solvent, and desorption time, were optimized, and the analytical performance of the developed SBSE-HPLC-UV method was evaluated under the optimized conditions. The limits of detection (LODs) of the proposed method for three NSAIDs were in the range of 0.23-0.31 μg L(-1), and the enrichment factors (EFs) were in the range of 51.6-56.3 (theoretical enrichment factor was 100). The reproducibility was also investigated at concentrations of 5, 20, and 100 μg L(-1), and the relative standard deviations (RSDs) were found to be less than 9.5, 7.5, and 7.6 %, respectively. The proposed method was successfully applied for the determination of NSAIDs in environmental water, urine, and milk samples.
Product Description
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O