Merck
  • Dihydromyricetin Protects against Bone Loss in Ovariectomized Mice by Suppressing Osteoclast Activity.
Dihydromyricetin Protects against Bone Loss in Ovariectomized Mice by Suppressing Osteoclast Activity.
Frontiers in pharmacology (2018-01-10)
Libo Zhao, Cong Cai, Jing Wang, Liming Zhao, Weijin Li, Changyu Liu, Hanfeng Guan, Yuanli Zhu, Jun Xiao
ABSTRACT
Dihydromyricetin (DMY), the main flavonoid component of
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)