Sign In to View Organizational & Contract Pricing
Select a Size
Product AMBH2D6F5728 is not currently sold in your country.
About This Item
Product AMBH2D6F5728 is not currently sold in your country.
Iupac Name
5,6,8,9-tetrahydro-7H-benzo[a]cyclohepten-7-one
Inchi Code
1S/C11H12O/c12-11-7-5-9-3-1-2-4-10(9)6-8-11/h1-4H,5-8H2
InChI key
MRLBDJHIJZHYMK-UHFFFAOYSA-N
storage temp.
Sealed in dry, store in freezer, under -20C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/37949-03-4.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/37949-03-4.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
