Skip to Content
Merck

860801P

Avanti

2-ACC

new

Avanti Research - A Croda Brand

Synonym(s):

2-ACC, 2-α-carboxy cholestanol

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C28H48O3
Molecular Weight:
432.68
UNSPSC Code:
12352211
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (860801P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CCC2C3CCC4C[C@@H](O)[C@@H](C[C@]4(C)C3CC[C@]12C)C(O)=O

General description

2-ACC is a synthetic Cholestanol; these compounds differ from cholesterol due to the lack of a double bond at the 5,6 position. Cholestanols have typically been used in the past to study the effects of sterols on liver function and cholesterol metabolism.

Application

Lipids Research

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service