Skip to Content
Merck

BDPH9BCCD53E

Bide Pharmatech

1-Heptanol

Synonym(s):

1-heptanol

Select a Size

Change View

About This Item

Linear Formula:
C7H16O1
CAS Number:
Molecular Weight:
116.2
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

Inchi Key

BBMCTIGTTCKYKF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: liquid Colour: colourless



Still not finding the right product?

Explore all of our products under 1-Heptanol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library