Skip to Content
Merck

ENAH95E73999

Enamine

2,4,5-tribromo-1-(difluoromethyl)-1H-imidazole

Sign In to View Organizational & Contract Pricing

Select a Size

Product ENAH95E73999 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
354.77
MDL number:
Product ENAH95E73999 is not currently sold in your country.

Iupac Name

2,4,5-tribromo-1-(difluoromethyl)-1H-imidazole

Inchi Code

1S/C4HBr3F2N2/c5-1-2(6)11(4(8)9)3(7)10-1/h4H

InChI key

VXYWJMIHCRGSIB-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-27152324

Pictograms

CorrosionExclamation mark

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-27152324


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library