Sign In to View Organizational & Contract Pricing
Select a Size
Product FLUH99CE4688 is not currently sold in your country.
Product FLUH99CE4688 is not currently sold in your country.
Iupac Name
1-(2-chloroethyl)-4-piperidinecarbonitrile hydrochloride
Inchi Code
1S/C8H13ClN2.ClH/c9-3-6-11-4-1-8(7-10)2-5-11;/h8H,1-6H2;1H
InChI key
DDMSKIKYADWREH-UHFFFAOYSA-N
storage temp.
Ambient Storage
Purity
98%
Country of Origin
GB
Product Link
https://fluorochem.co.uk/product/F924055
Signal Word
Warning
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/eng/sds_F924055_en.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
