Merck
EP6001099001
Eppendorf Fixation plate for stacking Racks
new
for use with CryoCube® F101h racks
Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
UNSPSC Code:
41103025
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
stainless steel
feature
to hold 2 x draw style CryoCube® F101h racks
for use with CryoCube® F101h racks
manufacturer/tradename
Eppendorf® 6001099001
packaging
pack of -1 ea
General description
Fixation plate to secure the top rack when stacking two stainless steel CryoCube F101 drawer racks.
Application
Securing freezer draw racks
Legal Information
CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)