Merck
  • α-Mangostin induces apoptosis and suppresses differentiation of 3T3-L1 cells via inhibiting fatty acid synthase.
α-Mangostin induces apoptosis and suppresses differentiation of 3T3-L1 cells via inhibiting fatty acid synthase.
PloS one (2012-03-20)
Xiaofang Quan, Yi Wang, Xiaofeng Ma, Yan Liang, Weixi Tian, Qingyun Ma, Hezhong Jiang, Youxing Zhao
ABSTRACT
α-Mangostin, isolated from the hulls of Garcinia mangostana L., was found to have in vitro cytotoxicity against 3T3-L1 cells as well as inhibiting fatty acid synthase (FAS, EC 2.3.1.85). Our studies showed that the cytotoxicity of α-mangostin with IC(50) value of 20 µM was incomplicated in apoptotic events including increase of cell membrane permeability, nuclear chromatin condensation and mitochondrial membrane potential (ΔΨm) loss. This cytotoxicity was accompanied by the reduction of FAS activity in cells and could be rescued by 50 µM or 100 µM exogenous palmitic acids, which suggested that the apoptosis of 3T3-L1 preadipocytes induced by α-mangostin was via inhibition of FAS. Futhermore, α-mangostin could suppress intracellular lipid accumulation in the differentiating adipocytes and stimulated lipolysis in mature adipocytes, which was also related to its inhibition of FAS. In addition, 3T3-L1 preadipocytes were more susceptible to the cytotoxic effect of α-mangostin than mature adipocytes. Further studies showed that α-mangostin inhibited FAS probably by stronger action on the ketoacyl synthase domain and weaker action on the acetyl/malonyl transferase domain. These findings suggested that α-mangostin might be useful for preventing or treating obesity.
Sigma-Aldrich
α-Mangostin, ≥98% (HPLC)
Pricing and availability is not currently available.