Merck
GETE70
GE TE70 ECL Transphor semi-dry transfer unit
gel size 14 cm (16 cm) , or 2 miniVue gels side by side, AC/DC input 100-240 V AC
ログインで組織・契約価格をご覧ください。
サイズを選択してください
表示を変更する
この商品について
NACRES:
NB.22
UNSPSC Code:
41105300
eCl@ss:
32160703
テクニカルサービス
お困りのことがあれば、経験豊富なテクニカルサービスチームがお客様をサポートします。
お手伝いします
packaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6210-34, Cytiva TE70
gel size
14 cm (16 cm) , or 2 miniVue gels side by side
General description
Designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amount of buffer. Units are available with and without built-in power supplies to give maximum flexibility.
  • •Electrotransfer proteins from polyacrylamide gels in less than 1 h in newly designed, state-of-the-art units.
  • •Tested for use with Amersham ECL Western blotting reagents
  • •Units are available with or without power supply.
  • •Use minimal buffer, only enough to saturate blotting papers and membranes.
  • •Long-lasting perforated platinum-titanium and stainless steel electrodes give uniform, contamination-free transfers.
  • •Transfer two polyacrylamide gels in a stack.
  • •Use low current and voltage for transfers.
最新バージョンのいずれかを選択してください:
試験成績書(COA)