Product Name
Decellularized ECM bioink precursor, from porcine skin, suitable for 3D bioprinting applications
form
semisolid
composition
Collagen (hydroxyproline), 90-125 μg/mg
GAG (biocolor), 0.4-0.8 μg/mg
color
white to beige
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
General description
Legal Information
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.
Protocols
Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.
Related Content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service