Skip to Content
Merck

DRAABPK-240K

MILLIPLEX® Dual Reporter Autoantibody IgG/IgM Reagent Pack

new

Companion pack to be used with MILLIPLEX® serology assays for detection of IgG and IgM antibodies

Synonym(s):

DR reagent pack, INTELLIFLEX dual reporter pack, IgG/IgM serotyping kit, Luminex® dual reporter serology assay, Millipore dual reporter autoantibody kit

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
41116126
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

input

plasma
serum

detection method

fluorometric (Luminex® xMAP® technology)

shipped in

wet ice

storage temp.

2-8°C

General description

Profiling both IgG and IgM autoantibodies in biological samples provides valuable insights into the immune response and potential disease conditions. While IgM autoantibodies are typically associated with a recent or concurrent immune response, IgG autoantibodies are suggestive of more chronic or persistent autoimmune processes.

Application

MILLIPLEX® Qualified assays undergo rigourous assay development, verification, and Quality Control testing to achieve optimal performance. Simultaneously measure antigen-specific IgG and IgM antibodies in human serum and plasma, when used in combination with MILLIPLEX® serology assays. The MILLIPLEX® Dual Reporter Autoantibody IgG/IgM Reagent Pack is a complementary add-on to any of the following MILLIPLEX® kits:

  • AAV Serotyping Panel (Cat. No. AAVSERG-230K )
  • Human Autoimmune Autoantibody Panel (Cat. No. HAIAB-10K)
  • Human Cancer Autoantibody Panel (Cat. No. HCABMAG-13K)
  • Human Neurodegenerative Autoantibody Panel (Cat. No. HNDGAB 220K)
  • Human Cytokine Autoantibody IgG Panel (Cat. No. HCYTABG-17K)
  • Human Cytokine Autoantibody IgM Panel (Cat. No. HCYTABM-17K)
Note: This product requires a xMAP® INTELLIFLEX DR-SE system

Assay Characteristics: Refer to kit protocol for assay precision.

Features and Benefits

  • Dual Reporter Capability: Specifically designed to utilize two lasers for simultaneous detection of both IgG and IgM serotypes for each analyte on a single bead.
  • Compatibility with Multiple MILLIPLEX® Kits: This reagent pack serves as a companion product to our expanding range of serology assays, enhancing versatility in testing.
  • Increased Data Throughput: Enables the measurement of more parameters per sample, saving time and resources.
  • Market-Exclusive Dual Reporter Assay: The only dual reporter serology assay available on the market, providing unique capabilities for simultaneous detection of IgG and IgM antibodies.
  • Easy-to-Follow Protocol: Streamlined and user-friendly protocols ensure quick setup and execution, minimizing training and reducing risk of errors in the laboratory.
  • Compatibility with the xMAP® INTELLIFLEX DRSE System: Specifically developed for use with the xMAP® INTELLIFLEX DRSE system, ensuring reliable and efficient assay results tailored for advanced multiplexing applications.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

For research use only. Not for use in diagnostic procedures.

Label License/Sticker for Assay Product: By opening the packaging containing this Assay Product (which contains fluorescently labeled microsphere beads authorized by Luminex Corporation) or using this Assay Product in any manner, you are consenting and agreeing to be bound by the End User Terms and Conditions and the End User License Agreement available at http://support.diasorin.com/end-user-terms-and-conditions/. If you do not agree to all of the terms and conditions, you must promptly return this Assay Product for a full refund prior to using it in any manner.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service