Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41116107
Skip To
grade
certified reference material
Agency
ERM®
form
solid
manufacturer/tradename
JRC
technique(s)
HPLC: suitable
ICP: suitable
gas chromatography (GC): suitable
application(s)
environmental
industrial qc
format
matrix material
storage temp.
2-8°C
Analysis Note
For more information please see: ERMCZ130
Legal Information
ERM is a registered trademark of European Commission
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service