Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Ships in 21 days - from Aldrich Partner.
| €645.64 |
5 mg | Ships in 21 days - from Aldrich Partner.
| €2,435.80 |
About This Item
Vendor SKU: AA00FBFG-1MG
€645.64
Inchi Code
1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,21H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+/m0/s1
Inchi Key
KJWZYMMLVHIVSU-IYCNHOCDSA-N
Purity
95%
Storage temp.
N/A
Country of Origin
US
1 of 1
This Item | |||
|---|---|---|---|
| description - | description A solution in methyl acetate | description A solution in ethanol at 100 µg/ml | description - |
| Purity 95% | Purity 98% | Purity 98% | Purity 99.95% |
| Storage temp. N/A | Storage temp. - | Storage temp. -20C | Storage temp. -80/-20 |
| Country of Origin US | Country of Origin US | Country of Origin US | Country of Origin CN |
| Inchi Code 1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,21H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+/m0/s1 | Inchi Code 1S/C20H30O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-17,21H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4+,13-12+/t15-,16+,17+/m0/s1 | Inchi Code 1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-18,21-22H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4+,13-12+/t15-,16+,17+,18-/m0/s1 | Inchi Code 1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-17,19,21,23H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4-,13-12+/t15-,16+,17+,19+/m0/s1 |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



