Skip to Content
Merck

Skip To

AABH97CD86E2

AA BLOCKS, INC.

Urocortin III (human) (trifluoroacetate salt)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Ships in 21 days - from Aldrich Partner.
€561.19

Vendor SKU: AA01EFJQ-1MG

€561.19


Ships in 21 days - from Aldrich Partner.

To order products, please contact your local dealer.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C185H307N53O50S2.C2HF3O2/c1-33-93(18)142(147(196)251)233-163(267)114(57-59-134(190)243)213-152(256)101(26)207-160(264)115(60-66-289-31)215-166(270)118(69-87(6)7)220-172(276)124(76-108-83-199-85-201-108)217-155(259)102(27)208-164(268)125(77-135(191)244)218-154(258)98(23)203-148(252)96(21)202-149(253)97(22)204-159(263)113(56-58-133(189)242)212-151(255)100(25)206-158(262)112(54-46-64-200-185(197)198)214-165(269)117(68-86(4)5)221-173(277)126(78-136(192)245)225-161(265)111(53-43-45-63-187)211-150(254)99(24)205-157(261)110(52-42-44-62-186)210-153(257)103(28)209-180(284)143(94(19)34-2)234-175(279)128(80-138(194)247)227-171(275)123(75-107-50-40-37-41-51-107)224-168(272)120(71-89(10)11)219-167(271)119(70-88(8)9)222-174(278)127(79-137(193)246)226-162(266)116(61-67-290-32)216-181(285)144(95(20)35-3)235-176(280)129(81-139(195)248)230-183(287)146(105(30)241)237-179(283)132-55-47-65-238(132)184(288)141(92(16)17)232-177(281)130(82-140(249)250)228-169(273)121(72-90(12)13)223-178(282)131(84-239)231-170(274)122(73-91(14)15)229-182(286)145(104(29)240)236-156(260)109(188)74-106-48-38-36-39-49-106;3-2(4,5)1(6)7/h36-41,48-51,83,85-105,109-132,141-146,239-241H,33-35,42-47,52-82,84,186-188H2,1-32H3,(H2,189,242)(H2,190,243)(H2,191,244)(H2,192,245)(H2,193,246)(H2,194,247)(H2,195,248)(H2,196,251)(H,199,201)(H,202,253)(H,203,252)(H,204,263)(H,205,261)(H,206,262)(H,207,264)(H,208,268)(H,209,284)(H,210,257)(H,211,254)(H,212,255)(H,213,256)(H,214,269)(H,215,270)(H,216,285)(H,217,259)(H,218,258)(H,219,271)(H,220,276)(H,221,277)(H,222,278)(H,223,282)(H,224,272)(H,225,265)(H,226,266)(H,227,275)(H,228,273)(H,229,286)(H,230,287)(H,231,274)(H,232,281)(H,233,267)(H,234,279)(H,235,280)(H,236,260)(H,237,283)(H,249,250)(H4,197,198,200);(H,6,7)/t93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104+,105+,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,127-,128-,129-,130-,131-,132-,141-,142-,143-,144-,145-,146-;/m0./s1

Inchi Key

ZMELYLRCZAEWRR-WABNFCLFSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library