Skip to Content
Merck

AABH97CE8FA2

AA BLOCKS, INC.

[2-(Acetyloxy)-5-nitro-1,3-phenylene]di(methylene) diacetate

Synonym(s):

2-(acetyloxy)-3-[(acetyloxy)methyl]-5-nitrobenzyl acetate

Select a Size

Change View

About This Item

Linear Formula:
C14H15O8N1
Molecular Weight:
325.28
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H15NO8/c1-8(16)21-6-11-4-13(15(19)20)5-12(7-22-9(2)17)14(11)23-10(3)18/h4-5H,6-7H2,1-3H3

Inchi Key

OTONBRJUYFCXLE-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library