Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 mg | Ships in 21 days - from Aldrich Partner.
| €223.37 |
250 mg | Ships in 21 days - from Aldrich Partner.
| €537.65 |
About This Item
Vendor SKU: AA003OKV-50MG
€223.37
Ships in 21 days - from Aldrich Partner.
Inchi Code
1S/2BH4.Ca/h2*1H4;/q2*-1;+2
Inchi Key
MOUPUCLGEWXWHJ-UHFFFAOYSA-N
Purity
97%
Storage temp.
RT
Country of Origin
US
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| Inchi Key MOUPUCLGEWXWHJ-UHFFFAOYSA-N | Inchi Key - | Inchi Key - | Inchi Key PRIILCVZLIFNKP-UHFFFAOYSA-N |
| Purity 97% | Purity - | Purity - | Purity - |
| Storage temp. RT | Storage temp. - | Storage temp. - | Storage temp. RT |
| Country of Origin US | Country of Origin - | Country of Origin - | Country of Origin US |
| Inchi Code 1S/2BH4.Ca/h2*1H4;/q2*-1;+2 | Inchi Code - | Inchi Code - | Inchi Code 1S/CH2N2.Ca.2H/c2-1-3;;;/h2H2;;; |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



