Skip to Content
Merck

Skip To

AABH9BD959CD

AA BLOCKS, INC.

N-Fmoc-N-methyl-3-methoxy-L-phenylalanine

Synonym(s):

(S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(3-methoxyphenyl)propanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C26H25O5N1
CAS Number:
Molecular Weight:
431.49
MDL number:
Product AABH9BD959CD is not currently sold in your country.


Inchi Code

1S/C26H25NO5/c1-27(24(25(28)29)15-17-8-7-9-18(14-17)31-2)26(30)32-16-23-21-12-5-3-10-19(21)20-11-4-6-13-22(20)23/h3-14,23-24H,15-16H2,1-2H3,(H,28,29)/t24-/m0/s1

Inchi Key

NRCFYIHAUKIDBL-DEOSSOPVSA-N

Purity

98%

Storage temp.

N/A

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library