Skip to Content

GF11368593

Copper-beryllium alloy, Cu98Be2

foil, 0.2m coil, 0.025mm thickness, as rolled

Synonym(s):

CU070280

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
31152304
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

foil (coil - as rolled)

composition

Cu98Be2

reaction suitability

core: beryllium, core: copper

manufacturer/tradename

Goodfellow 113-685-93

size × thickness

0.2 m × 0.025 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow


Still not finding the right product?

Explore all of our products under Copper-beryllium alloy, Cu98Be2




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library