Skip to Content
Merck

BDPH9BCC1DF0

Bide Pharmatech

4-Methyl-2-phenylthiazole-5-carboxylic acid

Synonym(s):

4-methyl-2-phenyl-1,3-thiazole-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H9O2N1S1
CAS Number:
Molecular Weight:
219.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H9NO2S/c1-7-9(11(13)14)15-10(12-7)8-5-3-2-4-6-8/h2-6H,1H3,(H,13,14)

Inchi Key

CRSMRBYEBHOYRM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to light-yellow powder or crystals



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library