Skip to Content
Merck

BDPH9BCC227B

Bide Pharmatech

3,5-Diphenyl-1H-pyrrole-2-carbaldehyde

Synonym(s):

3,5-diphenyl-1H-pyrrole-2-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H13O1N1
CAS Number:
Molecular Weight:
247.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H13NO/c19-12-17-15(13-7-3-1-4-8-13)11-16(18-17)14-9-5-2-6-10-14/h1-12,18H

Inchi Key

GRLGDALQUFILRJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library