Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C8H15N/c1-2-4-8(3-1)5-6-9-7-8/h9H,1-7H2
Inchi Key
NINJAJLCZUYDGV-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 2-Azaspiro[4.4]nonane
hstatements
Signal Word
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

