Skip to Content

BDPH9BCC4B5D

Bide Pharmatech

2-(Prop-2-yn-1-yl)pent-4-yn-1-ol

Synonym(s):

2-(prop-2-yn-1-yl)pent-4-yn-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10O1
CAS Number:
Molecular Weight:
122.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H10O/c1-3-5-8(7-9)6-4-2/h1-2,8-9H,5-7H2

Inchi Key

QCQDXFLZGCVLTR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 2-(Prop-2-yn-1-yl)pent-4-yn-1-ol


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library